Diacetyl, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2,3-Butanedione

M-Clarity™ quality level = MQ200

8.03528.0002 8.03528.0002
CA8.03528.0002EA 55.81 CAD
EM8.03528.0002 CA8.03528.0002
Diacetyl, Sigma-Aldrich®
Diacetyl
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.9904 g/cm³ (20 °C)
Flash Pt: 9 °C
MDL Number: MFCD00008756
CAS Number: 431-03-8
UN: 2346
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR