Lithium, Sigma-Aldrich®
Supplier: MilliporeSigma
8.05660.0010
8.05660.0010
CA8.05660.0010EA
160.44
CAD
CA8.05660.0010
EM8.05660.0010
Lithium, Sigma-Aldrich®
Lithium
Formula:
Li MW: 6.94 g/mol Boiling Pt: 1317 °C (1013 hPa) Melting Pt: 179 °C Density: 0.53 g/cm³ (20 °C) |
MDL Number:
MFCD00134051 CAS Number: 7439-93-2 UN: 1415 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...