Vileda® Professional CE Duo Mops, Vileda Professional-FHP
Supplier: Vileda Professional - FHP
141887 141886 147544 147545
89413-084CS 2798.81 CAD
89413-084 89413-086 10746-446 10746-448
Vileda® Professional CE Duo Mops, Vileda Professional-FHP
Mops
The less time spent on cleanroom cleaning, the less interference with keeping the core business running. The unique CE Duo Mop concept is the perfect answer for the high demands of effective cleaning and disinfection of cleanrooms and controlled environments. It is faster, easier, and twice as efficient as traditional methods. The CE Duo Mops allow users to achieve contact time requirements for common disinfectants, and still reach an excellent cleaning performance, with up to 99.99% bacteria removal. The CleanTech™ and SanTech™ Duo Mops form the most efficient and reliable cleaning concept in the market for contamination control.
  • Designed for use in cleanrooms up to close ISO 5/GMP A&B/FED209D Class 100
  • Adhesive-free lamination techniques are used to prevent additional product contamination
  • Suitable for bucket and pre-prepared method
  • Sterile items are validated to 10-6 SAL per ISO 11137
  • Manufactured and sterilized in the USA
  • Certificate of Sterility available upon request with each shipment
CleanTech™ Mops feature microfiber laminated cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
SanTech™ Mops feature polyester laminated cleanroom foam for disinfecting floors, walls, and ceilings. These mops offer excellent liquid absorbency. They are individually double bagged with Lot Number and Indicator Dot on each package.
Excellent microfiber cleaning results have been independently proven to provide bacteria reduction of 99.997%. CleanTech™ Mops are individually double bagged with Lot Number and Indicator Dot on each package.
Accessories information: The Swep Duo Plus Frame is extremely easy to use, and attaches to Vileda® extendable handles. After using one mop side, simply flip the frame over and continue cleaning with a clean mop. The lightweight and autoclavable frame features a removal lever that allow hands-free removal of used mops. All accessory handles are lightweight, ergonomic, and extremely durable. Handles extend with a twist of the wrist, and fit all Vileda® Professional frames, CE Squeegee, and other commonly used frames.