Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
1803-WF
89133-778CS 309.53 CAD
89133-778
Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Swabs Swabs for Cleaning
This specialty foam tipped applicator is produced with 100ppi reticulated medical grade foam
Made in the USA.
It has a soft, non-abrasive tip that is bonded securely to a white birch handle with thermoplastic adhesive.
Certifications: CE marked.