Quartz SUPRASIL® Macro and Micro Cells, PerkinElmer
Supplier: PerkinElmer
B0631011 B0631083 B0631082 B0631010 B0631077 B0631008 B0631007 B0631025 B0631008 B0631079 B0631080 B0631009 B0631079 B0631011 B0631071 B0631025 B0631071 B0631077 B0631081 B0631029 B0631012 B0631010 B0631049 B0631049 B0631027 B0631027 B0631009 B0631026 B0631012 B0631030
97012-874EA 3650.04 CAD
97012-874 CA97012-884 45603-698 97012-872 45603-810 97012-878 45603-736 45603-812 CA97012-882 45603-814 CA97013-524 CA97013-546 97012-870 97013-544 97013-524 97013-546 97012-924 CA97012-878 97012-884 CA97012-874 45603-742 97012-882 45603-744 CA97012-870 CA97012-872 45603-808 CA97013-536 CA97013-544 97013-536 CA97012-924
Quartz SUPRASIL® Macro and Micro Cells, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Standard rectangular macro and micro cells are the most frequently used type of spectroscopy cell for routine liquids analysis.
  • Clear windows for easy observation
  • Widely used in spectroscopy procedures
  • Rectangular design
Quartz SUPRASIL® cells are available in a range of sizes for use with various sample volumes. All cells contain two clear windows.