NIR Quartz SUPRASIL® 300 Macro Cells, PerkinElmer
Supplier: Perkin Elmer
B0631033 B0631013 B0631015 B0631035 B0631017 B0631018 B0631034 B0631016 B0631014 B0631036 B0631032 B0631015
CA97013-534PK 817.11 CAD
45603-710 45603-754 45603-720 45603-712 45603-756 97013-534 CA97013-534 45603-716 45603-748 45603-718 45603-750 45603-752
NIR Quartz SUPRASIL® 300 Macro Cells, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Standard rectangular macro cells are the most frequently used type of spectroscopy cell for routine liquids analysis.
  • Clear windows for easy observation
  • Widely used in spectroscopy procedures
  • Rectangular design
NIR quartz SUPRASIL® 300 cells are available in a range of sizes for use with various sample volumes. All macro cells contain two clear windows.