Two-Position Snap Caps, Simport Scientific
Supplier: Simport Scientific
T401-3DSPE T401-3DSPP T401-10DSPP T401-10DSPE
CA60829-031CS 130.7 CAD
CA60829-031 CA60872-807 CA60872-808 CA73960-344
Two-Position Snap Caps, Simport Scientific
Closures Test/Sample Tube Closures
Dual position caps are suitable for use in both aerobic and anaerobic procedures.
  • Cap for aerobic and anaerobic procedures
  • For use with 12 and 17 mm tubes
  • Polystyrene and polypropylene options available
Caps offer two possibilities. The closed but unsealed position where samples are maintained aerobic for microbiological procedures; and the fully sealed position where the cap is pushed tight to seal the tube for anaerobic use or for storage, transfer and centrifuge applications. Not autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable