Compact Titrators, METTLER TOLEDO
Supplier: Mettler Toledo
30267114 30267115 30252660 30252661 30267116 30252666 30267117 30252669 30252671
10861-674EA 15297.5 CAD
10861-674 10861-676 10861-678 10861-684 10861-686 10861-688 10861-690 10861-692 10861-694 75853-862
Compact Titrators, METTLER TOLEDO
Titrators
METTLER TOLEDO Compact Titrators are designed for routine applications and simple operation.
The large color touchscreen with the One Click user interface is easy to understand and operate. The touchscreen is optimized for direct access to routine tasks and provides clear information to the user.
Security is an important concept in all METTLER TOLEDO Titrators. Many parameters are automatically checked by the instrument, allowing you to keep full control of the titration analysis. Connectivity to LabX Titration Software allows for security and efficient management of results.