CELLview™ Slides, Greiner Bio-One
Supplier: Greiner Bio-One
543079 543979
10836-545CS 1307.96 CAD
10836-545 10836-587
CELLview™ Slides, Greiner Bio-One
Slides Microscope Slides
For the Cultivation, Staining, and Microscopy of Cells.
  • 10 wells with alphanumeric coding
  • Detachable black compartmentalisation
  • 96 well microtiter plate design
  • Reduced meniscus effect
  • Positioning notch for automated microscopy
  • Highly transparent achromatic borosilicate glass, hydrolytic class 1
CELLview™ slide has been developed in collaboration with the EMBL, Heidelberg and consists of a transparent slide with an embedded cover glass and a black detachable compartmentalisation
CELLview™ slide is excellently suited for all microscopic applicationsMore Product Information