VWR® VividPro™ Fluorescent Stain
Supplier: VWR
VividPro™ is a generic fluorescent stain for protein gel electrophoresis that can be used to visualize proteins.
- Protein gel electrophoresis
- Intermediate sensitivity between ruthenium based dyes and Coomassie stains
- Compatible with mass spectrometry analysis
VividPro™ is a generic fluorescent stain for protein gel electrophoresis that can be used to visualize proteins in SDS PAGE gels with a sensitivity that is intermediate between ruthenium based dyes and Coomassie stains
VividPro™ (0.2 mL or 0.5 mL) is supplied as a 10,000X staining concentrate.
Visualization can be achieved following the electrophoretic run using a fast staining protocol, and is compatible with mass spectrometry analysis.