Platinum Tipped Tongs, Heraeus
Supplier: HERAEUS MATERIALS TECHNOLOGY NA
GE20/230 GE20/300 GE20/400 GE20/500 GE20/600
75797-122EA 4000.09 CAD
75797-122 75797-124 75797-126 75798-806 75798-808
Platinum Tipped Tongs, Heraeus
Tongs
These conventional crucible tongs have been fitted with 99.95% Pure Platinum Shoes and are available in various lengths.
Although Platinum Crucibles and Dishes are made to withstand extreme temperatures and are resistant to many acids and corrosive media, they can be damaged due to careless handling.
It is therefore recommended that Platinum Crucibles and/or Dishes be handled with Platinum Tipped Tongs while placing in and removing from a hot furnace.