Polysoft Face Shields, OP-D-OP
Supplier: OP-D-OP INC
206PVNY 206PVRB 206PVMG
33000-456EA 142.2 CAD
33000-456 33000-454 33000-452
Polysoft Face Shields, OP-D-OP
Face Shields
Face shield is designed for facial protection in medical facilities and laboratories.
  • Unbreakable polycarbonate shield can be replaced
  • Cinch cord can be adjusted for a secure fit
  • Crystal clear optics
  • Polysoft band for comfort
Fits over glasses and replaces goggles and masks.