3" Test Sieves, W.S. Tyler™
Supplier: W.S. Tyler
1300 1306 1310 1290 1292 1293 1294 1295 1296 1297 1298 1299 1301 1302 1303 1304 1305 1307 1308 1542 1544 1545 1547 1549 1551 1557 1553 1539 1540 1541 1543 1546 1548 1555 1559
57321-945EA 240.52 CAD
57321-945 57321-950 30620-686 30624-134 30624-136 30624-138 30624-140 30624-142 30624-144 30624-146 30624-148 30624-150 30624-152 30624-154 30624-156 30624-158 30624-160 30624-162 30624-164 30624-166 30624-168 30624-170 30624-172 30624-174 30624-176 30624-178 30621-290 89423-898 89424-000 89424-002 89424-004 89424-006 89424-008 89424-010 89424-012
3" Test Sieves, W.S. Tyler™
Sieves Test Sieves
These test sieves are available with stainless or brass frame and stainless cloth.
  • Depth to cloth: 1-1/4"