Corning® Ice Pan and Buckets, Corning
Supplier: Corning
Corning®
432105 432102 432099 432135 432095 432122 432097 432133 432132 432134 432114 432101 432106 432107 432117 432100 432121 432094 432093 432131 432124 432103 432113 432123 432126 432120 432098 432116 432127 432110 432112 432111 432109 432115 432096 432130 432128 432108 432118 432129 432119 432104 432125
75779-980CS 129.67 CAD
75779-980 75779-914 75779-958 75779-936 75779-912 75779-934 75779-978 75779-956 75779-918 75779-916 75779-938 75779-950 75779-972 75779-970 75779-910 75779-932 75779-954 75779-976 75779-952 75779-974 75779-930 75779-904 75779-926 75779-948 75779-804 75779-924 75779-946 75779-902 75779-968 75779-908 75779-928 75779-906 75779-962 75779-984 75779-940 75779-982 75779-960 75779-988 75779-966 75779-922 75779-986 75779-920 75779-964
Corning® Ice Pan and Buckets, Corning
Ice Containers
Ethylene vinyl acetate (EVA) foam.
  • Solid, one piece moulded, unbreakable
  • Resistant to highly acidic and basic solutions, salts, alcohol, acetone, chloroform, phenol, etc.
  • Insulative properties and resistant to temperatures from −196 to 93 °C
Ideal for use with ice, dry ice, liquid nitrogen, saline solution and acetone.
Corning ice pans and buckets are multi-purpose pans and buckets that are a great non-toxic alternative to conventional ice pans.
EVA has little or no odor, is inert and used in biomedical engineering applications such as drug delivery devices. The multi-purpose containers are highly insulative, stackable, unbreakable, lightweight and leak-proof. Containers will not 'sweat' or deform and have a textured finish on the bottom to prevent slipping on the benchtop. Ideal for use with ice, dry ice (−78℃), liquid nitrogen (−196℃), alcohol or saline solutions, or warm solutions (+93°C). Available in five sizes and seven vibrant colors.