Rigid-Lock QUICKBERM®, Justrite®
Supplier: Justrite
28518 28522 28528
10861-140EA 2320.95 CAD
10861-140 10861-144 10861-150
Rigid-Lock QUICKBERM®, Justrite®
Spill Control Materials
Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic, and cleaning.
  • Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup - no assembly required
  • Durable EnGuard™V, modified PVC coated fabric withstands long-term UV exposure
Exceptionally strong structural integrity includes a reinforced CriticalCorner™ design featuring wraparound diagonal radio frequency welds to prevent billowing,More Product Information