Maintenance Spill Berm, Justrite®
Supplier: Justrite
28404 28412 28421 28424
10861-178EA 940.62 CAD
10861-178 10861-186 10861-194 10861-198
Maintenance Spill Berm, Justrite®
Spill Control Materials
Compact and flexible spill containment quickly contain minor spills and leaks during temporary storage of drums, machine parts, batteries, and equipment.
  • No assembly required - simply unfold and use
  • Encased urethane foam sidewalls offer easy roll-through access without the need for a ramp
  • Walls quickly spring back into place with no memory loss
Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner. Anchor stakes not included. Order optional grates to keepMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable