VWR® Plus Series Freestanding Countertop and Undercounter Freezers
Supplier: VWR
VWR®
HCUCFS0440 HCUCFS0120ALH HCUCFS0120 HCUCFS0120LH HCUCFS0430 HCUCFS0120A
76210-392EA 2586.62 CAD
76210-392 10819-892 10819-662 10834-034 10819-664 76210-390
VWR® Plus Series Freestanding Countertop and Undercounter Freezers
Freezers Laboratory / General Purpose Freezers
These undercounter and countertop freezers provide lab quality cold storage in a space-saving design.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft circulation
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Select models offer auto defrost cycles for convenient defrosting. For lower temperatures, -30 and -40 °C models are available.
Certifications: *Select models ENERGY STAR certified: 10819-662, 10819-664, 76210-392, 76210-390, 10819-892
Ordering information: All units have a two-year parts and labor warranty. Select models come with an additional three-year compressor parts warranty. (Please refer to unit datasheets to confirm).