Corning® XT Starter Ice-free Cooler, Corning
Supplier: Corning
432014 432015
75779-828CS 390.46 CAD
75779-828 75779-830
Corning® XT Starter Ice-free Cooler, Corning
Coolers Benchtop Coolers
An open-platform ice-free cooler that accommodates most CoolRack® and CoolSink® modules.
  • Consists of a base, cooling core and a lid
  • Includes a 1 to 8 °C temperature indicator for visual assurance of proper temperature
  • To use with most Corning CoolRack® and CoolSink® Modules
  • The internal cooling or freezing cores provide 0.5 to 4 °C for up to 4 hours
  • One module capacity
  • Purple color
Low profile and small footprint make it ideal for use in the hood, keeping samples cold (0.5 to 4.0°C) up to four hours. 1 to 8 °C temperature indicator provides visualMore Product Information