VWR® Plus Series Undercounter Refrigerators Freestanding with Natural Refrigerants
Supplier: VWR International
10819-646 10819-870 10819-872 10819-876 10834-032 76210-102 76210-104 76300-508
10819-646EA 1607.21 CAD
10819-646 10819-870 10819-872 10819-876 10834-032 76210-102 76210-104 76300-508
VWR® Plus Series Undercounter Refrigerators Freestanding with Natural Refrigerants
Refrigerators
These undercounter and countertop refrigerators provide lab quality cold storage in a space-saving design.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft circulation
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency. Glass door and left-hinged models are available.
Certifications: *Select models ENERGY STARMore Product Information