Lysing Matrix A, MP Biomedicals
Supplier: MP Biomedicals
116910500 116910050 116910100 116910050 116910100
IC116910500EA 2935.34 CAD
IC116910500 75784-610 75784-612 CA76046-060 CA76046-062
Lysing Matrix A, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA.
  • Chemically inert
  • Will not bind nucleic acids
  • Used for all sample types except soil
Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA
Each impact-resistant 2 mL tube contains garnet matrix and one, 1/4 inch ceramic sphere. Extra 1/4 inch ceramic spheres are packaged separately. Lysing Matrix A tubes have orange caps and are found in the FastDNA™ and FastRNA™ SPIN Kits. Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA, RNA, functional proteins and other metabolites and small molecules. This grinding matrix is chemically inert and will not bind nucleic acids.
This grinding matrix is chemically inert and will not bind nucleic acids.