FastRNA™, Pro Kit, MP Biomedicals
Supplier: MP Biomedicals
FastRNA™
116035050 116045050 116025050 116070050
75784-594EA 583.4 CAD
75784-594 75784-588 75784-592 75784-590
FastRNA™, Pro Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastRNA™ Pro Kit isolates total RNA for RT-PCR analysis and other downstream applications
  • Pro Blue Kit is designed to efficiently isolate total RNA from gram positive and gram negative bacteria
  • Pro Red Kit is designed to efficiently isolate total RNA from yeast and fungi
  • Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells
  • Pro Soil-Direct Kit is designed to efficiently isolate total RNA from organic material found in soil samples
The FastRNA™ Pro Blue Kit is designed to efficiently isolate total RNA from gram positive and gram negative bacteria
The FastRNA™ Pro Red Kit is designed to efficiently isolate total RNA from yeast and fungi. Using the FastPrep-24 or FastPrep FP120 Instrument, up to 10⁹ cells are homogenized by Lysing Matrix C in impact-resistant 2 mL tubes. Total RNA is released into the proprietary, protective RNApro™Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
The FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells. Using the FastPrep-24 or FastPrep FP120 Instrument, between 50-500 mg of tissue can be homogenized by Lysing Matrix D in impact-resistant 2 mL tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
The FastRNA™ Pro Soil-Direct Kit is designed to efficiently isolate total RNA from organic material found in soil samples. Using any FastPrep® Instrument, up to 0.5 g of material is homogenized by Lysing Matrix E in impact-resistant 2 mL tubes. Total RNA is released into a protective solution, extracted, precipitated, and cleaned with the proprietary RNAMATRIX™ and a Quick-Clean Spin Filter. Total RNA prepared with this kit is suitable for RT-PCR analysis and other downstream applications. Isolation of total RNA directly from soil samples.