Vial Rack for Headspace Vials, Restek
Supplier: Restek
22643 22644
10854-122EA 69.09 CAD
10854-122 10854-124
Vial Rack for Headspace Vials, Restek
Racks Cryogenic Vial Racks
These vial racks securely holds up to 36 headspace vials.
  • Autoclavable up to 121°C for 20 minutes
  • Vials lock in rack for convenient one-handed cap removal
  • Easy to clean and fully autoclavable
  • Stackable even with vials in place
The openings in the bottom of each well facilitate drainage and the alphanumeric indexing on rack simplifies sample identification.