Defined PressFit Connectors, Restek
Supplier: Restek
22160 22162 22164 22166 22168 22170
10852-916EA 598.32 CAD
10852-916 10852-922 10852-924 10852-936 10852-938 10852-940
Defined PressFit Connectors, Restek
Fittings and Connectors
PressFit connectors feature ID-specific ends for better sealability and a defined fit.
  • Deactivated borosilicate glass
  • Connect a guard column to an analytical column
  • Repair a broken column
  • Connect a column outlet to a transfer line
These connectors are color-coded for easy identification.