Extended Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Supplier: Restek
22633 22634 22635 22636 22637
10856-036EA 245.92 CAD
10856-036 10856-038 10856-040 10856-042 10856-044
Extended Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Fittings and Connectors
Visible below inlet insulating cup for easy, breakage-free column installation without removing cup.
  • Finger-tight design to allow wrench-free use
  • Available for standard (1/16") and compact (Agilent-style) ferrules
  • Made from high-quality stainless steel
  • Meets or exceeds original manufacturer's performance