Click-On In-Line Super Clean® Big Trap Replacement Filters, Restek
Supplier: Restek
27205 27204 27206
10854-852EA 1647.81 CAD
10854-852 10854-894 10855-440
Click-On In-Line Super Clean® Big Trap Replacement Filters, Restek
Filters Gas Filters
Big Trap filters are effective at trapping oxygen, water, and hydrocarbons to help protect against instrument damage.
  • 2.0 L oxygen capacity
  • 120 g total hydrocarbons
  • 70 g water
Ordering information: Note: Replacement filter only—fittings sold separately. See Click-On in-line Super Clean® Big Trap filter kits for more information.