Click-On In-Line Super Clean® Big Trap Filter Kits, Restek
Supplier: Restek
27203 27202 27201
10855-352EA 1823.87 CAD
10855-352 10855-578 10855-598
Click-On In-Line Super Clean® Big Trap Filter Kits, Restek
Filters Gas Filters
'Quick connect' fittings with spring-loaded check valves allow filter replacement in seconds without tools or contamination.
  • Largest-capacity traps available—run longer without replacing
  • Stainless steel construction withstands pressures up to 1000 psig
  • High-purity output ensures 99.9999% pure gas (maximum flow of 8 L/min)
Triple gas trap is ideal for purifying carrier gas.
Hydrocarbon trap removes hydrocarbon contamination from carrier gas to help eliminate ghost peaks.