Capillary Column Rinsing Reservoir, Reservoir only, Restek
Supplier: Restek
20612 20613
RK20612EA 345.84 CAD
RK20612 RK20613
Capillary Column Rinsing Reservoir, Reservoir only, Restek
Reagent Reservoirs
Rinsing reservoirs can be filled with solvent rinses and are ideal for cleaning capillary columns.
Restore the performance of bonded-phase capillary columns by dissolving and removing soluble, nonvolatile residue using this reservoir kit.
The 50 mL rinsing reservoir is equipped with 1/4 " inlet and outlet connections and includes a built-in fritted disk to prevent particulate matter from contaminating the column. The kit includes reservoir, pressure regulator, fittings, ferrules, and tubing. Reservoir also available separately.
Delivery information: Column not included.