Viton® O-Rings for Agilent GCs, Restek
Supplier: Restek
22241 22242
10058-746PK 36 CAD
10058-746 10803-278
Viton® O-Rings for Agilent GCs, Restek
Seals Sealing Rings
Viton® O-rings are for use with a number of Agilent GC systems.
  • Available in packs of 10 or 50 depending on model
  • Fit split (6.3 mm OD) or splitless (6.5 mm OD) liners
For use with Agilent GC systems.