Standard Ferrules for PerkinElmer GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20231 20212 27064 27065
RK20231PK 442.54 CAD
RK20231 RK20212 10852-526 10852-528
Standard Ferrules for PerkinElmer GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Standard ferrules fit ID MXT® columns with internal diameters of 0.25 mm or 0.32 mm
  • VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability
  • VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Stable to 400°C
  • Recommended for mass spec transfer lines
For use with PerkinElmer GCs.