Standard Ferrules for PerkinElmer GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Supplier: Restek
20211 20229 27062 27063
RK20211PK 120.23 CAD
RK20211 RK20229 10803-414 10852-508
Standard Ferrules for PerkinElmer GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Fittings and Connectors
Standard ferrules fit fused silica columns with internal diameters ≤0.28 mm.
  • VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability
  • VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Stable to 400°C
  • Recommended for mass spec transfer lines