SPI with Buffer for Bruker/Varian GCs equipped with SPI inlets, Restek
Supplier: Restek
20851
RK20851PK 425.24 CAD
RK20851
SPI with Buffer for Bruker/Varian GCs equipped with SPI inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Borosilicate glass fabrication
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.