Merlin Microseal™ Septa for Shimadzu GCs, Restek
Supplier: Restek
22972
10855-840EA 855.99 CAD
10855-840
Merlin Microseal™ Septa for Shimadzu GCs, Restek
Septa
A Merlin Microseal septum provides several distinct advantages: elimination of septum coring, long lifetime, and consistently low needle-insertion force.
  • Compatible with Shimadzu models GC-2010 and GC-2025
  • 450 °C maximum injection port temperature
  • For use with 23-gauge (0.63 mm, 0.025") needle or probe with blunt, truncated conical tip
The Microseal septum incorporates two separate sealing mechanisms; these sliding seals prevent septum coring and the associated accumulation of septum crumbs in the injection port liner. Installation is simple, requiring no modification of the injection port.
Delivery information: Includes Nut, Adapter, O-Ring, and 2 General-Purpose (#410) Microseals.