Interlock Vials, Restek
Supplier: Restek
22433 22435 22439 22434 22436 22437 22438 22440
10058-772PK 249.34 CAD
10058-772 10058-774 10058-776 10854-106 10854-108 10854-110 10854-112 10854-114
Interlock Vials, Restek
Vials
Interlock limited volume vials offer the performance and convenience of a one-piece microvial for a fraction of the cost.
  • Clear or amber color
  • Screw-thread, crimp-top, or snap-ring formats
  • Glass fabrication
A 300 µl insert is fused into a 12×32 mm clear or amber glass vial. This one-piece design reduces the potential for contamination.