Flip Seal® Dual Vespel® Ring Inlet Seals, Restek
Supplier: Restek
23413 23406 23411 23412 23414
10803-200EA 556.13 CAD
10803-200 10856-120 10856-122 10856-124 10856-126
Flip Seal® Dual Vespel® Ring Inlet Seals, Restek
Seals Mechanical Seals
The reversible, two-sided design of these ring inlet seals allow significantly more analyses than other seals.
  • Vespel ring embedded in top and bottom surface eliminates need for a washer
  • Very little torque required to make seal
Highly inert gold or Siltek®-deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column.
Ordering information: Note: The Flip Seal® inlet seal requires a special reducing nut adaptor fitting, which is included in the kit.
The Flip Seal® Adaptor can be used with standard 1/16" ferrules.