Ferrules for SRI & Buck Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20255 20205 27080
RK20255PK 85 CAD
RK20255 RK20205 10852-532
Ferrules for SRI & Buck Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Fits columns with internal diameters of 0.25 mm or 0.32 mm
  • Durable, leak-tight
  • Preconditioned to eliminate out-gassing
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines
For use with SRI and Buck Scientific GCs.