Ferrules for Thermo Scientific GCs, Fit 0.45/0.53 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20285 20284
10057-904EA 280.62 CAD
10057-904 10852-554
Ferrules for Thermo Scientific GCs, Fit 0.45/0.53 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
For use with Thermo Scientific GCs
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite
  • Stable to 450°C
These ferrules contain no binders that can off-gas or adsorb analytes. Suitable for columns with internal diameters of 0.45 - 0.53 mm.