Ferrules for Thermo Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20283 20282
10057-902EA 186.4 CAD
10057-902 10852-552
Ferrules for Thermo Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
For use with Thermo Scientific GCs
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite
  • Stable to 450°C
These ferrules contain no binders that can off-gas or adsorb analytes. Suitable for columns with internal diameters of 0.25 mm or 0.32 mm.