Ferrules for Thermo Scientific GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Supplier: Restek
20280 20281
RK20280PK 67.71 CAD
RK20280 RK20281
Ferrules for Thermo Scientific GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Fittings and Connectors
For use with Thermo Scientific GCs
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite
  • Stable to 450°C
These ferrules contain no binders that can off-gas or adsorb analytes. Suitable for columns with internal diameters of 0.18–0.28 mm.