Standard Ferrules, Fit 0.53 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20202 20224 22216
RK20202PK 103.34 CAD
RK20202 RK20224 10852-608
Standard Ferrules, Fit 0.53 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Standard ferrules are ideal for use with MXT® GCs.
  • Durable, leak-tight
  • Preconditioned to eliminate out-gassing
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines
Ferrules feature internal diameters of 0.8 mm, compatible with columns 0.45 or 0.53 mm in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable