Compact Ferrules for Agilent GCs, Fit 0.53 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20263 20252 20253 20264 27074 27075
RK20263PK 157.54 CAD
RK20263 RK20252 RK20253 RK20264 10803-418 10852-550
Compact Ferrules for Agilent GCs, Fit 0.53 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Compact ferrules are ideal for use with Agilent GCs.
  • Durable, leak-tight
  • Preconditioned to eliminate out-gassing
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines
Ferrules feature internal diameters of 0.8 mm, compatible with columns 0.45 or 0.53 mm in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable