Standard Ferrules, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Supplier: Restek
20200 20227 22214
RK20200PK 98.38 CAD
RK20200 RK20227 10852-604
Standard Ferrules, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Fittings and Connectors
Standard ferrules fit fused silica columns with internal diameters ≤0.28 mm.
  • Durable, leak-tight
  • Preconditioned to eliminate out-gassing
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines