Parker Balston® Nitrogen Gas Generators for LC-MS, Restek
Supplier: Restek
22131
10058-726EA 40665.09 CAD
10058-726
Parker Balston® Nitrogen Gas Generators for LC-MS, Restek
Gas Generators Nitrogen Generators
This nitrogen gas generators turn compressed air into ultra-pure nitrogen (up to 99.5%).
  • Flows from 1 to 44 L/min
  • Models N2-04, N2-14, N2-22, and N2-35 require no electricity
  • Safe and reliable
  • Low maintenance
  • Maintenance kits include replacement filters
Accessories information: For use with Parker Balston® model FID-1000 and FID-2500 gas stations and Filters should be changed every year, resin and desiccant cartridges can be replaced every 2 years.