Toggle Valve, Parker Balston® Shut-Off Gas Valve, Restek
Supplier: Restek
22190 22191 22188 22189
10855-712EA 456.55 CAD
10855-712 10855-714 10855-774 10855-776
Toggle Valve, Parker Balston® Shut-Off Gas Valve, Restek
Valves Air Regulation Valves
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
Toggle valves provide 1/4 turn for on-off control of fluids utilized in process and instrumentation applications. Available with connection sizes of 1/8" or 1/4" in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable