Union, Swagelok®, Restek
Supplier: Restek
23115 23116 23117 23165 23166 23167 22540 22575 22541 22542 22577 22909
10058-890PK 231.16 CAD
10058-890 10058-892 10058-894 10058-934 10058-936 10058-938 10855-432 10855-434 10855-490 10855-492 10855-502 10855-504 10855-632 10855-786
Union, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/16", 1/8", 1/4", or 3/8" in diameter.