Port Connector, Swagelok®, Restek
Supplier: Avantor
23132 22559 22557 22558 22593 23182 23180 23181
10803-326PK 111.06 CAD
10803-326 10803-328 10854-622 10854-654 10854-656 10854-874 10855-026 10855-028 10855-178 10855-474 10855-476 10855-614
Port Connector, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/8" or 1/4" in diameter.