Male Connector, Swagelok®, Restek
Supplier: Restek
23133 23134 23183 23185 22563 23184 23186 23187 22562 22564 23136 23137 22610 22596 22612 22561 22565 22611
10058-910PK 292.05 CAD
10058-910 10058-912 10058-956 10854-472 10854-480 10854-482 10854-484 10854-486 10854-632 10854-634 10854-772 10854-774 10854-776 10854-876 10854-880 10854-882 10854-884 10854-906 10854-908 10854-912 10855-052 10855-530
Male Connector, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available in 1/16" to 1/4" NPT sizing.