Cross, Swagelok®, Restek
Supplier: Restek
23124 23174 22551 22552 22586 23175
10058-906PK 303.77 CAD
10058-906 10058-908 10058-952 10855-498 10855-500 10855-510 10855-512 10855-656
Cross, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Use these cross connectors for making four-way connections with tubing. Available with connection sizes of 1/8" or 1/4" in diameter.