Bulkhead Union, Swagelok®, Restek
Supplier: Restek
22570 22571 22617 22618
10855-018EA 260.28 CAD
10855-018 10855-020 10855-030 10855-032
Bulkhead Union, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/8" or 1/4" in diameter.