Tube End Reducer, Parker, Restek
Supplier: Restek
21544 21545 21546 21547 21933 21934
10058-696PK 77.42 CAD
10058-696 10854-514 10854-516 10854-538 10854-640 10854-642 10854-648 10855-118 10855-120 10855-228 10855-230 10855-436 10855-438 10855-514 10855-516
Tube End Reducer, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/16", 1/8", or 1/4" in diameter.