Tee, Parker, Restek
Supplier: Restek
21827 21828 21926 21927 21928 22526 22527 22528
RK21827PK 208.14 CAD
RK21827 10855-648 10855-700 10855-752 10855-770 10855-772 10855-790 10855-810 10855-812 10855-814 10855-824 10855-826
Tee, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Use these tee connectors for making three-way connections with tubing. Available with connection sizes of 1/16", 1/8", or 1/4" in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable